3-Fluoro-4-isopropoxy-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

46206-500MG 46206-1G 46206-5G
77338-208EA 469.68 USD
77338-208 77350-692 77374-942
3-Fluoro-4-isopropoxy-5-(trifluoromethyl)benzoic acid ≥97%
3-Fluoro-4-isopropoxy-5-(trifluoromethyl)benzoic acid

Caution: for Research Use Only

Formula: C₁₁H₁₀F₄O₃
Storage Temperature: Ambient
MDL Number: MFCD33403821
CAS Number: 2624417-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR