2-Benzyloxy-5-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Benzyloxypyridine-3-boronic acid pinacol ester, 6-Benzyloxypyridine-3-boronic acid, pinacol ester, 2-(Benzyloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

38868-1G 38868-5G 38868-25G
77340-668EA 60.77 USD
77340-668 77363-148 77374-692
2-Benzyloxy-5-pyridineboronic acid pinacol ester ≥97%
2-Benzyloxy-5-pyridineboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₈H₂₂BNO₃
Storage Temperature: Ambient
MDL Number: MFCD06798263
CAS Number: 832735-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR