4-Bromo-2,6-difluoro-N,N-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

38234-500MG 38234-1G 38234-5G 38234-25G
77326-646EA 139.05 USD
77326-646 77344-662 77362-938 77385-726
4-Bromo-2,6-difluoro-N,N-dimethylbenzamide ≥97%
4-Bromo-2,6-difluoro-N,N-dimethylbenzamide

Caution: for Research Use Only

Formula: C₉H₈BrF₂NO
Storage Temperature: Ambient
MDL Number: MFCD23840436
CAS Number: 1515527-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR