(2-(Benzyloxy)-3,4-difluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

37893-500MG 37893-1G 37893-5G
77336-350EA 387.28 USD
77336-350 77350-200 77374-400
(2-(Benzyloxy)-3,4-difluorophenyl)methanol ≥97%
(2-(Benzyloxy)-3,4-difluorophenyl)methanol

Caution: for Research Use Only

Formula: C₁₄H₁₂F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD32707046
CAS Number: 2156187-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR