1-Chloro-4-ethoxy-2,3-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

37460-500MG 37460-1G 37460-5G 37460-25G
77326-390EA 232.78 USD
77326-390 77350-082 77362-702 77385-540
1-Chloro-4-ethoxy-2,3-difluorobenzene ≥95%
1-Chloro-4-ethoxy-2,3-difluorobenzene

Caution: for Research Use Only

Formula: C₈H₇ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD28750528
CAS Number: 1384466-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR