2-(5,6-Dimethylpyridin-3-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

35263-500MG 35263-1G 35263-5G
77325-808EA 333.72 USD
77325-808 77349-474 77373-714
2-(5,6-Dimethylpyridin-3-yl)propan-2-ol ≥95%
2-(5,6-Dimethylpyridin-3-yl)propan-2-ol

Caution: for Research Use Only

Formula: C₁₀H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD31805982
CAS Number: 1862885-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR