Methyl 3-cyano-2,4-difluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

33754-500MG 33754-1G 33754-5G
77337-744EA 440.84 USD
77337-744 77349-248 77373-506
Methyl 3-cyano-2,4-difluorobenzoate ≥95%
Methyl 3-cyano-2,4-difluorobenzoate

Caution: for Research Use Only

Formula: C₉H₅F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD28758892
CAS Number: 1805664-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR