2-Bromo-5-iodophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31096-500MG 31096-1G 31096-5G 31096-25G
77325-204EA 99.91 USD
77325-204 77342-766 77361-552 77373-230
2-Bromo-5-iodophenylboronic acid pinacol ester ≥97%
2-Bromo-5-iodophenylboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₂H₁₅BBrIO₂
Storage Temperature: Ambient
MDL Number: MFCD29088257
CAS Number: 2121512-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR