1-(4-Bromophenyl)-4-methylcyclohexanol ≥97%

Supplier: AOBChem USA

Warning

28615-500MG 28615-1G 28615-5G
77338-592EA 503.67 USD
77338-592 77348-772 77373-034
1-(4-Bromophenyl)-4-methylcyclohexanol ≥97%
1-(4-Bromophenyl)-4-methylcyclohexanol

Caution: for Research Use Only

Formula: C₁₃H₁₇BrO
Storage Temperature: Ambient
CAS Number: 87625-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR