6-Chloro-2-fluoro-3-iodobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

26047-500MG 26047-1G 26047-5G 26047-25G
77324-752EA 176.13 USD
77324-752 77346-512 77361-076 77384-396
6-Chloro-2-fluoro-3-iodobenzyl alcohol ≥97%
6-Chloro-2-fluoro-3-iodobenzyl alcohol

Caution: for Research Use Only

Formula: C₇H₅ClFIO
Storage Temperature: Ambient
MDL Number: MFCD24848716
CAS Number: 1449008-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR