4-[(4-Bromo-3-chlorophenyl)methyl]-morpholine hydrochloride ≥97%

Supplier: AOBChem USA

Warning

24838-500MG 24838-1G 24838-5G 24838-25G
77324-588EA 199.82 USD
77324-588 77348-434 77360-920 77384-272
4-[(4-Bromo-3-chlorophenyl)methyl]-morpholine hydrochloride ≥97%
4-[(4-Bromo-3-chlorophenyl)methyl]-morpholine hydrochloride

Caution: for Research Use Only

Formula: C₁₁H₁₄BrCl₂NO
Storage Temperature: Ambient
MDL Number: MFCD22628225
CAS Number: 952290-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR