1,3-Dichloro-2-isopropoxybenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
24413-1G
24413-5G
24413-25G
77341-498EA
90.64
USD
77341-498
77360-868
77372-676
1,3-Dichloro-2-isopropoxybenzene ≥97%
1,3-Dichloro-2-isopropoxybenzene
Caution: for Research Use Only
Formula:
C₉H₁₀Cl₂O Storage Temperature: Ambient |
MDL Number:
MFCD22543697 CAS Number: 57678-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...