2-Benzyloxy-3-fluorophenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-(2-(Benzyloxy)-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Benzyloxy-3-fluorobenzeneboronic acid pinacol ester
23386-500MG
23386-1G
23386-5G
23386-25G
77324-438EA
206
USD
77324-438
77348-316
77360-770
77384-154
2-Benzyloxy-3-fluorophenylboronic acid pinacol ester ≥97%
2-Benzyloxy-3-fluorophenylboronic acid pinacol ester
Caution: for Research Use Only
Formula:
C₁₉H₂₂BFO₃ Storage Temperature: Ambient |
MDL Number:
MFCD23098817 CAS Number: 1661043-58-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...