6-(Methoxymethoxy)naphthalene-2-boronic acid ≥97%

Supplier: AOBChem USA

22018-500MG 22018-1G 22018-5G 22018-25G
77324-364EA 231.75 USD
77324-364 77348-258 77360-688 77384-094
6-(Methoxymethoxy)naphthalene-2-boronic acid ≥97%
6-(Methoxymethoxy)naphthalene-2-boronic acid

Caution: for Research Use Only

Formula: C₁₂H₁₃BO₄
MDL Number: MFCD21000128
CAS Number: 2246782-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR