(3-Oxo-2,3-dihydro-1H-inden-5-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

18261-500MG 18261-1G 18261-5G 18261-25G
77324-160EA 250.29 USD
77324-160 77348-122 77360-424 77383-946
(3-Oxo-2,3-dihydro-1H-inden-5-yl)boronic acid ≥97%
(3-Oxo-2,3-dihydro-1H-inden-5-yl)boronic acid

Caution: for Research Use Only

Formula: C₉H₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD22191954
CAS Number: 1135871-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR