1,3-Dibromo-2-ethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

17775-1G 17775-5G 17775-25G
77340-594EA 60.77 USD
77340-594 77360-402 77372-336
1,3-Dibromo-2-ethoxybenzene ≥95%
1,3-Dibromo-2-ethoxybenzene

Caution: for Research Use Only

Formula: C₈H₈Br₂O
Storage Temperature: Ambient
MDL Number: MFCD31805979
CAS Number: 38751-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR