4-Chloro-2,5-difluoropyridine ≥95%
Supplier: AOBChem USA
|
Warning
|
13967-500MG
13967-1G
13967-5G
13967-25G
77323-912EA
129.78
USD
77323-912
77344-294
77360-104
77372-068
4-Chloro-2,5-difluoropyridine ≥95%
4-Chloro-2,5-difluoropyridine
Caution: for Research Use Only
Formula:
C₅H₂ClF₂N Storage Temperature: Ambient |
MDL Number:
MFCD09031178 CAS Number: 851386-40-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...