2-Chloro-1-(6-Chloro-2,3-difluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

117739-500MG 117739-1G 117739-25G 117739-5G
77323-610EA 284.28 USD
77323-610 77347-688 77383-574 77371-730
2-Chloro-1-(6-Chloro-2,3-difluorophenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(6-Chloro-2,3-difluorophenyl)-2,2-difluoroethanone

Caution: for Research Use Only

Formula: C₈H₂Cl₂F₄O
Storage Temperature: Ambient
CAS Number: 1879995-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR