5,6-Dichloro-N-isopropylnicotinamide ≥95%

Supplier: AOBChem USA

Warning

113402-500MG 113402-1G 113402-5G 113402-25G
77323-508EA 191.58 USD
77323-508 77347-580 77359-622 77383-500
5,6-Dichloro-N-isopropylnicotinamide ≥95%
5,6-Dichloro-N-isopropylnicotinamide

Caution: for Research Use Only

Formula: C₉H₁₀Cl₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD09813244
CAS Number: 345582-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR