1-(tert-Butyl)-4-fluorobenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
109820-1G
109820-5G
109820-25G
77341-896EA
106.09
USD
77341-896
77359-570
77371-538
1-(tert-Butyl)-4-fluorobenzene ≥97%
1-(tert-Butyl)-4-fluorobenzene
Caution: for Research Use Only
Formula:
C₁₀H₁₃F Storage Temperature: Ambient |
MDL Number:
MFCD15143585 CAS Number: 701-30-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...