3-Chloro-N,N-dimethyl-5-(trifluoromethyl)-2-pyridinamine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-N,N-dimethyl-5-(trifluoromethyl)pyridin-2-amine

107389-500MG 107389-1G 107389-5G 107389-25G
77323-414EA 158.62 USD
77323-414 77345-748 77359-510 77383-444
3-Chloro-N,N-dimethyl-5-(trifluoromethyl)-2-pyridinamine ≥97%
3-Chloro-N,N-dimethyl-5-(trifluoromethyl)-2-pyridinamine

Caution: for Research Use Only

Formula: C₈H₈ClF₃N₂
Storage Temperature: Ambient
MDL Number: MFCD13561831
CAS Number: 157071-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR