1-(3,5-Dichloro-2-fluorophenyl)-2-fluoroethanone ≥97%

Supplier: AOBChem USA

Warning

106410-500MG 106410-1G 106410-5G 106410-25G
77323-398EA 263.68 USD
77323-398 77347-460 77359-494 77383-432
1-(3,5-Dichloro-2-fluorophenyl)-2-fluoroethanone ≥97%
1-(3,5-Dichloro-2-fluorophenyl)-2-fluoroethanone

Caution: for Research Use Only

Formula: C₈H₄Cl₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD34779010
CAS Number: 2299889-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR