3-Bromo-2-chloro-6-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

101390-500MG 101390-1G 101390-5G 101390-25G
77323-310EA 190.55 USD
77323-310 77347-386 77359-392 77383-366
3-Bromo-2-chloro-6-hydroxybenzaldehyde ≥97%
3-Bromo-2-chloro-6-hydroxybenzaldehyde

Caution: for Research Use Only

Formula: C₇H₄BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD23705600
CAS Number: 1427373-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR