4-(4-Benzyloxy-5-bromo-2-pyrimidinyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Benzyloxy-5-bromo-2-morpholin-4-yl-pyrimidine

10136-500MG 10136-1G 10136-5G 10136-25G
77323-308EA 90.64 USD
77323-308 77342-412 77359-390 77371-380
4-(4-Benzyloxy-5-bromo-2-pyrimidinyl)morpholine ≥97%
4-(4-Benzyloxy-5-bromo-2-pyrimidinyl)morpholine

Caution: for Research Use Only

Formula: C₁₅H₁₆BrN₃O₂
MW: 350.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798240
CAS Number: 885952-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR