6-Fluoro-5-propoxypyridin-3-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
100615-500MG
100615-1G
100615-5G
100615-25G
77323-296EA
281.19
USD
77323-296
77347-374
77359-374
77383-358
6-Fluoro-5-propoxypyridin-3-ol ≥97%
6-Fluoro-5-propoxypyridin-3-ol
Caution: for Research Use Only
Formula:
C₈H₁₀FNO₂ MW: 171.169 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34776394 CAS Number: 2901095-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...