Drug Affected Baby Simulators
Supplier: American 3B Scientific
3004302 3004306 3004307 3004402 3014303
470352-880EA 33.4 USD
470352-880 470352-888 470352-874 470352-868 470352-898 470352-872 470352-876 470352-896
Drug Affected Baby Simulators
Models Human Biology Models
The first manikin to simulate a baby's varying needs is even better.
  • Simulates either drug prevention or teen pregnancy prevention
  • Programmed tending events that are unpredictable to the student, but easy for the teacher to monitor
  • Cries, coos, and burps in response to appropriate care
  • Programs are 48 hours in length and run continuously
Every manikin is both a drug prevention baby and a general teen pregnancy prevention baby with four programs. Teach students how to properly support a baby’s head with a bendable neck feature, how to differentiate between shaken baby syndrome and other types of abuse.
Programing provides a teacher-activated day care pause when needed during the school day. Includes three different tending programs typical of most babies, to simulate normal baby tending needs along with fussy periods in each program. There are 25 to 27 tending occurrences in each 48 hour program. A fourth tending program simulates the more annoying cry of a drug- or alcohol-affected baby. The simulator requires care 31 times instead of the standard 25, in a 48-hour period. The manikin also continues to cry 30 seconds longer after being tended by the student. Providing a teacher with complete control of the childcare experience. The model includes a 'panic' key for quieting difficulties without discontinuing the entire simulation and alerts the teacher to a student’s abuse, panic, or tampering with the control box. Comes complete with one set of student keys, one set of teacher keys, reproducible parental consent form/permission slip, reproducible student response sheet, teacher correction template, diaper, 9 V battery, and instructions.