The Belly Bath, 230 V, IBI Scientific
Supplier: IBI Scientific
BWBAA220V
89408-320EA 5176.69 USD
89408-320
The Belly Bath, 230 V, IBI Scientific
Baths Shaking Water Baths
The IBI Belly Bath is a versatile laboratory instrument
  • 230 V model, not for use in USA
The Belly Bath combines a water bath with the gentle agitation of the Belly Dancer making it suitable for numerous Life Science applications. The Belly Bath is also an ideal unit for multiple uses as the Belly Dancer can be used for other applications with the water bath removed.
Applications for the Belly Bath include Nucleic Acid Hybridizations, Over-Night Lysis of Tissue Samples, Northern and Southern Filter Blot Hybridizations, High-Stringency Filter Washes, In-Situ Hybridizations and Washes, and Hot Staining Protocols and Recombinant DNA Technology.
The light-weight, fast-heating water bath can be placed on the Belly Dancer platform or run separately on the side of the unit. The microprocessor PID controller has a set point control pad and digital display of actual water temperature. The control panel on the Belly Dancer contains a dedicated, illuminated ON/OFF switch for the water bath. The water bath incorporates thermal run-away safety switch, which operates independently of the main temperature controller. The lid is 3/6” thick PET clear lid with an easy grip handle and thermometer port.
Certifications: This 230V unit is CE marked.
Caution: 230 V model, not for use in USA. Corresponding model with voltage suitable for use in US (89408-318) is also available, please see separate product page.