Probe-based qPCR 2X master mix, G-Biosciences
Supplier: G-Biosciences
786-1982
786-1983
786-1984
76716-652EA
81.21
USD
76716-652
76716-656
76716-704
Probe-based qPCR 2X master mix, G-Biosciences
Nucleic Acid Reagents
qPCR/RT-qPCR Enzymes and Kits
Probe-based qPCR 2x master mix contains Taq DNA polymerase, low ROX reference dye, dNTPs and optimized buffer with stabilizers. This master mix is ideal for high-throughput real-time PCR screening and validation.
- Enhanced efficiency, specificity, and sensitivity
- Compatible with all real-time PCR instruments
- Superior gene expression results under various cycling conditions
Applications:
Gene expression analysis
Genotyping
Mutation detection, pathogen detection, viral detection and absolute quantification
Caution: Stable for one year when stored at -20 °C.