VWR® PerfectBlue Twin L Dual Gel System
Supplier: VWR
VWR®
85-2020WCS
700-1080EA 1756.31 USD
700-1080
VWR® PerfectBlue Twin L Dual Gel System
Electrophoresis Systems Vertical Gel Electrophoresis Systems
The largest of the series, the Twin L can accommodate 20×20 cm gels for the simultaneous separation of up to 50 samples making this system ideally suited to either higher throughput or separating lower molecular weight samples. The system actively manages temperature build up with an efficient water cooling system and central aluminium ceramic plates for effective and safe long-term operation.
*Based on an average gel and a tooth at 20 mm depth
Ordering information: Complete system, with or without casting base, includes internal cooling core, SuperSafe cover with attached leads, 4× glass plate pair sets, 4× combs (1,5 mm thick, 2×15 teeth and 2×20 teeth), 4× spacer sets (1,5 mm), 2× replacement gaskets, 1× blocking plate (for single gel runs) and 1× SpacerPlate.