IncuFridge Refrigerated Incubators
Supplier: Revolutionary Science
RS-IF-328P RS-IF-328S RS-IF-365P
470353-538EA 3028.9 USD
470353-538 470353-540 470353-542
IncuFridge Refrigerated Incubators
Incubators Cooled Incubators
IncuFridge refrigerated chilling incubators – ideal for heating, cooling, and maintaining temperature.
  • Compact design
  • Easy calibration
  • Refrigerant-free, Peltier based
  • Thick, closed cell insulation
  • Rapid ramp rate to set point
The Incufridge 300 series is the reliable and affordable option for your heating and cooling needs. These refrigerated incubators (Peltier-based, refrigerant-free) are the ultimate in flexibility when refrigerating or incubating temperature sensitive material. Each 300 series Incufridge is thermostatically controlled and has aMore Product Information
Order Now
SPECIFICATIONS
Model IncuFridge Pro 28L model IncuFridge Standard model IncuFridge Pro 65L model
Environmentally Preferable
Int. W×D×H 30.5× 35.6×25.7 cm (12×14×10.125") 30.5× 35.6×25.7 cm (12×14×10.125") 41×41×38.4 cm (16.125×16.125×15.125")
Ext. W×D×H 38.7×59.1×42.5 cm (15.25×23.25×16.75") 38.7×59.1×42.5 cm (15.25×23.25×16.75") 51.8×63.5×57.5 cm (20.375×25×22.625")