Anti-ACTL7B Goat Polyclonal Antibody

Supplier: Antibodies.com LLC
A84436-100
77068-472EA 480 USD
77068-472
Anti-ACTL7B Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to ACTL7B for ELISA and WB with samples derived from Human.

  • Validated Applications: ELISA, W

Type: Primary
Antigen: ACTL7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR