Anti-DOCK1 Goat Polyclonal Antibody

Supplier: Antibodies.com LLC
A83785-100
77069-358EA 494.4 USD
77069-358
Anti-DOCK1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to DOCK1 for WB with samples derived from Human.

  • Validated Applications: W

Type: Primary
Antigen: DOCK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR