Anti-KIR3DL3 Rabbit Polyclonal Antibody

Supplier: Antibodies.com LLC
A8518-50 A8518-100
76864-428EA 225 USD
76864-428 76850-648
Anti-KIR3DL3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to KIR3DL3 for WB and IHC with samples derived from Human.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:20

Type: Primary
Antigen: KIR3DL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR