Anti-Factor I Mouse Monoclonal Antibody [clone: KT22]

Supplier: Antibodies.com LLC
A24-200 A24-100
77041-710EA 480 USD
77041-710 77041-712
Anti-Factor I Mouse Monoclonal Antibody [clone: KT22]
Antibodies
Mouse monoclonal (KT22) antibody to Factor I for ELISA with samples derived from Human.

  • Validated Applications: ELISA
  • Recommended Dilutions: ELISA: 1 µg/ml

Type: Primary
Antigen: Factor I
Clonality: Monoclonal
Clone: KT22
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR