Anti-Calmegin Rabbit Monoclonal Antibody [clone: ARC2214]

Supplier: Antibodies.com LLC
A305871-100 A305871-50
77219-796EA 305 USD
77219-796 77219-794
Anti-Calmegin Rabbit Monoclonal Antibody [clone: ARC2214]
Antibodies
Rabbit monoclonal [ARC2214] antibody to Calmegin for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC, ICC/IF
  • Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: Calmegin
Clonality: Monoclonal
Clone: ARC2214
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR