Anti-LRG1 Mouse Monoclonal Antibody [clone: LRG1/4882]

Supplier: Antibodies.com LLC
A277544-100
76965-944EA 540.75 USD
76965-944
Anti-LRG1 Mouse Monoclonal Antibody [clone: LRG1/4882]
Antibodies
Mouse monoclonal [LRG1/4882] antibody to LRG1 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: LRG1
Clonality: Monoclonal
Clone: LRG1/4882
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR