Anti-NAPSIN A Rabbit Recombinant Antibody [clone: NAPSA/1865R]

Supplier: Antibodies.com LLC
A253855-100
76954-910EA 540.75 USD
76954-910
Anti-NAPSIN A Rabbit Recombinant Antibody [clone: NAPSA/1865R]
Antibodies
Recombinant rabbit monoclonal [NAPSA/1865R] antibody to NAPSIN A for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: NAPSIN A
Clonality: Recombinant
Clone: NAPSA/1865R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR