Anti-TRIM24 Goat Polyclonal Antibody

Supplier: Antibodies.com LLC
A83286-100
77065-760EA 494.4 USD
77065-760
Anti-TRIM24 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to TRIM24 for ELISA and IHC with samples derived from Human.

  • Validated Applications: ELISA, IHC

Type: Primary
Antigen: TRIM24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR