Anti-ARHGEF18 Goat Polyclonal Antibody

Supplier: Antibodies.com LLC
A82796-100
77075-288EA 480 USD
77075-288
Anti-ARHGEF18 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to ARHGEF18 for ELISA and WB with samples derived from Human.

  • Validated Applications: ELISA, WB

Type: Primary
Antigen: ARHGEF18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR