Anti-Neurofascin Goat Polyclonal Antibody

Supplier: Antibodies.com LLC
A82546-100
77071-194EA 480 USD
77071-194
Anti-Neurofascin Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Neurofascin for ELISA and WB with samples derived from Human.

  • Validated Applications: ELISA, WB

Type: Primary
Antigen: Neurofascin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR