Anti-HRASLS2 Goat Polyclonal Antibody

Supplier: Antibodies.com LLC
A82467-100
77072-412EA 480 USD
77072-412
Anti-HRASLS2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to HRASLS2 for ELISA and IHC with samples derived from Human.

  • Validated Applications: ELISA, IHC

Type: Primary
Antigen: HRASLS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR