Anti-CD30 Mouse Recombinant Antibody [clone: rKi-1/779]

Supplier: Antibodies.com LLC
A250652-100
76964-422EA 525 USD
76964-422
Anti-CD30 Mouse Recombinant Antibody [clone: rKi-1/779]
Antibodies
Recombinant mouse monoclonal [rKi-1/779] antibody to CD30 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD30
Clonality: Recombinant
Clone: rKi-1/779
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR