Anti-TCL1 Mouse Monoclonal Antibody [clone: TCL1/2079]

Supplier: Antibodies.com LLC
A250397-100
76951-654EA 525 USD
76951-654
Anti-TCL1 Mouse Monoclonal Antibody [clone: TCL1/2079]
Antibodies
Mouse monoclonal [TCL1/2079] antibody to TCL1 for WB and IHC-P with samples derived from Human.

  • Validated Applications: WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: TCL1
Clonality: Monoclonal
Clone: TCL1/2079
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR