Anti-Cytochrome C Mouse Monoclonal Antibody [clone: 7H8.2C12]

Supplier: Antibodies.com LLC
A249643-100
76955-520EA 525 USD
76955-520
Anti-Cytochrome C Mouse Monoclonal Antibody [clone: 7H8.2C12]
Antibodies
Mouse monoclonal [7H8.2C12] antibody to Cytochrome C for WB, Flow Cytometry and IHC-P with samples derived from Human, Mouse, Rat, Horse, Canine, Pigeon, Frog and Drosophila.

  • Validated Applications: WB, Flow Cytometry, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, Flow Cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cytochrome C
Clonality: Monoclonal
Clone: 7H8.2C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human;Mouse;Rat;Horse;Dog;Pigeon;Frog;Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR