Anti-CD23 Mouse Monoclonal Antibody [clone: FCER2/3592]

Supplier: Antibodies.com LLC
A248529-100
76949-694EA 525 USD
76949-694
Anti-CD23 Mouse Monoclonal Antibody [clone: FCER2/3592]
Antibodies
Mouse monoclonal [FCER2/3592] antibody to CD23 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: FCER2/3592
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR