Anti-CD45 Mouse Monoclonal Antibody [clone: MEM-28] (PE-Cyanine 7)

Supplier: Antibodies.com LLC
A122014-100
76963-672EA 470 USD
76963-672
Anti-CD45 Mouse Monoclonal Antibody [clone: MEM-28] (PE-Cyanine 7)
Antibodies
Mouse monoclonal [MEM-28] antibody to CD45 (PE-Cyanine 7) for Flow Cytometry with samples derived from Human.

  • Validated Applications: Flow Cytometry
  • Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: MEM-28
Conjugation: PE-Cyanine 7
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR