Anti-CD3 epsilon Mouse Monoclonal Antibody [clone: APA1/1] (APC (Allophycocyanin))

Supplier: Antibodies.com LLC
A121842-100
76958-392EA 442.9 USD
76958-392
Anti-CD3 epsilon Mouse Monoclonal Antibody [clone: APA1/1] (APC (Allophycocyanin))
Antibodies
Mouse monoclonal [APA1/1] antibody to CD3 epsilon (APC) for Flow Cytometry with samples derived from Mouse and Human.

  • Validated Applications: Flow Cytometry
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/ml

Type: Primary
Antigen: CD3 epsilon
Clonality: Monoclonal
Clone: APA1/1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR