Anti-Pig IgG Fc Goat Polyclonal Antibody (AP (Alkaline Phosphatase) Then)

Supplier: Antibodies.com LLC
A294482-500
76833-752EA 283.25 USD
76833-752
Anti-Pig IgG Fc Goat Polyclonal Antibody (AP (Alkaline Phosphatase) Then)
Antibodies
Goat anti-pig IgG Fc (AP) secondary antibody for WB, IHC, ICC and ELISA with samples derived from Pig.

  • Validated Applications: WB, IHC, ICC, ELISA
  • Recommended Dilutions: ELISA: 1:5,000 to 1:50,000, ICC: 1:100 to 1:1,000, IHC: 1:100 to 1:1,000, WB: 1:5,000 to 1:50,000

Type: Secondary
Antigen: Pig IgG Fc
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR