Anti-HYAL1 Goat Polyclonal Antibody

Supplier: Antibodies.com LLC
A285977-100
76958-370EA 494.4 USD
76958-370
Anti-HYAL1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to HYAL1 for ELISA, IHC, IF and Flow Cytometry with samples derived from Human.

  • Validated Applications: ELISA, IHC, IF, Flow Cytometry

Type: Primary
Antigen: HYAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR