Anti-p53 Rabbit Polyclonal Antibody

Supplier: Antibodies.com LLC
A95522-100
77050-636EA 473.8 USD
77050-636
Anti-p53 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to p53 (phospho Ser9) for WB, IHC and ELISA with samples derived from Human.

  • Validated Applications: WB, IHC, ELISA
  • Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:10000

Type: Primary
Antigen: p53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR